Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATCAGGCTGTCCTTTCTTTCC[A/G]TAAGCCCATTCTGGTTCAATCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186947 MIM: 614907 | ||||||||||||||||||||
Literature Links: |
FKBP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FKBP3 - FK506 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002013.3 | 616 | Silent Mutation | TAC,TAT | Y,Y 198 | NP_002004.1 | |
XM_011536565.2 | 616 | Silent Mutation | TAC,TAT | Y,Y 165 | XP_011534867.1 | |
XM_017021092.1 | 616 | Silent Mutation | TAC,TAT | Y,Y 198 | XP_016876581.1 | |
XM_017021093.1 | 616 | Silent Mutation | TAC,TAT | Y,Y 165 | XP_016876582.1 |
PRPF39 - pre-mRNA processing factor 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD127 - small nucleolar RNA, C/D box 127 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |