Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGATGGCCGAGTGCTTCACCTTG[C/G]CCTTCTGGGCAGCGCTTAAGCCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606167 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR132 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR132 - G protein-coupled receptor 132 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278694.1 | 851 | Missense Mutation | GCC,GGC | A,G 238 | NP_001265623.1 | |
NM_001278695.1 | 851 | Missense Mutation | GCC,GGC | A,G 229 | NP_001265624.1 | |
NM_001278696.1 | 851 | Missense Mutation | GCC,GGC | A,G 50 | NP_001265625.1 | |
NM_013345.3 | 851 | Missense Mutation | GCC,GGC | A,G 238 | NP_037477.1 |