Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCAGGCAGCTCACTGACCCTTC[A/G]CATCGGAACAGGCCCAGCTACGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601686 | ||||||||||||||||||||
Literature Links: |
TEP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEP1 - telomerase associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319035.1 | 6304 | Silent Mutation | TGC,TGT | C,C 2485 | NP_001305964.1 | |
NM_007110.4 | 6304 | Silent Mutation | TGC,TGT | C,C 2593 | NP_009041.2 | |
XM_005268027.4 | 6304 | Silent Mutation | TGC,TGT | C,C 2593 | XP_005268084.1 | |
XM_011537110.2 | 6304 | Silent Mutation | TGC,TGT | C,C 2070 | XP_011535412.1 |