Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGGTCATGGTCCTGGAGGTGG[C/T]AAGAAGGATGACAAGGTAAATATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602706 | ||||||||||||||||||||
Literature Links: |
PSMC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMC1 - proteasome 26S subunit, ATPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002802.2 | 1609 | Intron | NP_002793.2 | |||
XM_017021470.1 | 1609 | UTR 5 | XP_016876959.1 | |||
XM_017021471.1 | 1609 | Intron | XP_016876960.1 |