Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATTTGGAACAGGCTGTTGTTGA[C/T]GTGATCTCTGTGGTCCCAAACCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609486 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EAPP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EAPP - E2F associated phosphoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318916.1 | 887 | Missense Mutation | ATC,GTC | I,V 128 | NP_001305845.1 | |
NM_018453.3 | 887 | Missense Mutation | CAT,CGT | H,R 167 | NP_060923.2 | |
XM_011536958.1 | 887 | Missense Mutation | CAT,CGT | H,R 103 | XP_011535260.1 | |
XM_017021460.1 | 887 | Missense Mutation | CAT,CGT | H,R 167 | XP_016876949.1 |