Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAACGAGGTACCTGCAGTTCGGC[C/T]CGTTCCACCTCCCAGTGCGCCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607243 MIM: 614766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP4S1 - adaptor related protein complex 4 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128126.2 | 486 | Intron | NP_001121598.1 | |||
NM_001254726.1 | 486 | Intron | NP_001241655.1 | |||
NM_001254727.1 | 486 | Intron | NP_001241656.1 | |||
NM_001254728.1 | 486 | Intron | NP_001241657.1 | |||
NM_001254729.1 | 486 | Intron | NP_001241658.1 | |||
NM_007077.4 | 486 | Intron | NP_009008.2 | |||
XM_005267293.4 | 486 | Intron | XP_005267350.1 | |||
XM_011536371.2 | 486 | Intron | XP_011534673.1 | |||
XM_011536372.2 | 486 | Intron | XP_011534674.1 |
STRN3 - striatin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083893.1 | 486 | Silent Mutation | CGA,CGG | R,R 90 | NP_001077362.1 | |
NM_014574.3 | 486 | Silent Mutation | CGA,CGG | R,R 90 | NP_055389.3 | |
XM_005267569.3 | 486 | Silent Mutation | CGA,CGG | R,R 90 | XP_005267626.1 | |
XM_005267570.3 | 486 | Silent Mutation | CGA,CGG | R,R 90 | XP_005267627.1 |