Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTACCTTGGGGGATTCCCACAGC[A/G]CTTCCAACAGGGCTTCTCTGGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604562 MIM: 611137 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACIN1 - apoptotic chromatin condensation inducer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDH24 - cadherin 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022478.3 | 1718 | Silent Mutation | AGC,AGT | S,S 486 | NP_071923.2 | |
NM_144985.3 | 1718 | Intron | NP_659422.2 | |||
XM_011537089.1 | 1718 | Silent Mutation | AGC,AGT | S,S 486 | XP_011535391.1 |
LOC105370705 - collagen alpha-2(I) chain-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMB11 - proteasome subunit beta 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |