Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAAGTTCCAGGCCACATCTGAG[C/T]GGCGCACAGGGCTCCAAATGATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608577 MIM: 138319 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHURC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHURC1 - churchill domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHURC1-FNTB - CHURC1-FNTB readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202558.1 | 619 | Intron | NP_001189487.1 | |||
NM_001202559.1 | 619 | Intron | NP_001189488.1 |
GPX2 - glutathione peroxidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002083.3 | 619 | Missense Mutation | CAC,CGC | H,R 147 | NP_002074.2 |
RAB15 - RAB15, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |