Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGATACTGTACCTAGGAGCACA[C/G]GGATGCCTGTATTGGGCCTTCTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607203 | ||||||||||||||||||||
Literature Links: |
SAV1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAV1 - salvador family WW domain containing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021818.3 | 1474 | Silent Mutation | CCC,CCG | P,P 265 | NP_068590.1 | |
XM_011537057.2 | 1474 | Silent Mutation | CCC,CCG | P,P 232 | XP_011535359.1 | |
XM_017021568.1 | 1474 | Silent Mutation | CCC,CCG | P,P 232 | XP_016877057.1 |