Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600654 MIM: 602161 MIM: 612487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMC9 - ER membrane protein complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR7703 - microRNA 7703 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME1 - proteasome activator subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME2 - proteasome activator subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002818.2 | 410 | Intron | NP_002809.2 | |||
XM_006720213.1 | 410 | Intron | XP_006720276.1 |
RNF31 - ring finger protein 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310332.1 | 410 | Intron | NP_001297261.1 | |||
NM_017999.4 | 410 | Missense Mutation | GCA,GTA | A,V 54 | NP_060469.4 |