Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGGAACTTCAATATCCTCTTC[C/T]TTCTCTTCATCCTCCTCAGATCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606241 | ||||||||||||||||||||
Literature Links: |
DICER1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DICER1 - dicer 1, ribonuclease III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195573.1 | 5515 | Intron | NP_001182502.1 | |||
NM_001271282.2 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | NP_001258211.1 | |
NM_001291628.1 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | NP_001278557.1 | |
NM_030621.4 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | NP_085124.2 | |
NM_177438.2 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | NP_803187.1 | |
XM_011536599.1 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | XP_011534901.1 | |
XM_011536600.2 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | XP_011534902.1 | |
XM_011536601.2 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | XP_011534903.1 | |
XM_011536602.2 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | XP_011534904.1 | |
XM_011536604.2 | 5515 | Silent Mutation | AAA,AAG | K,K 1663 | XP_011534906.1 | |
XM_011536605.2 | 5515 | Silent Mutation | AAA,AAG | K,K 1305 | XP_011534907.1 | |
XM_017021120.1 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | XP_016876609.1 | |
XM_017021121.1 | 5515 | Silent Mutation | AAA,AAG | K,K 1798 | XP_016876610.1 | |
XM_017021122.1 | 5515 | Silent Mutation | AAA,AAG | K,K 1663 | XP_016876611.1 | |
XM_017021123.1 | 5515 | Silent Mutation | AAA,AAG | K,K 1663 | XP_016876612.1 |