Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCGGACTATCTTCGCCAGCCAC[A/G]GCAACTTCTTCAACATGTGAGTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613317 MIM: 612028 MIM: 600654 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF11 - DDB1 and CUL4 associated factor 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EMC9 - ER membrane protein complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FITM1 - fat storage inducing transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203402.2 | 348 | Missense Mutation | AGC,GGC | S,G 84 | NP_981947.1 |
PSME1 - proteasome activator subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |