Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCTTCACCATTCTGCATCTCCA[A/G]TGGCTTTGGAAAATATATAATCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2A1 - general transcription factor IIA subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278940.1 | 1613 | Missense Mutation | ACT,ATT | T,I 321 | NP_001265869.1 | |
NM_015859.3 | 1613 | Missense Mutation | ACT,ATT | T,I 371 | NP_056943.1 | |
NM_201595.2 | 1613 | Missense Mutation | ACT,ATT | T,I 332 | NP_963889.1 |