Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCACCTGATAACAACCTTTTCTG[C/G]ATTTATTACTGTCTCTTTCCACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TC2N PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TC2N - tandem C2 domains, nuclear | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128595.2 | 1641 | Missense Mutation | CCA,GCA | P,A 477 | NP_001122067.1 | |
NM_001128596.2 | 1641 | Missense Mutation | CCA,GCA | P,A 477 | NP_001122068.1 | |
NM_001289134.1 | 1641 | Missense Mutation | CCA,GCA | P,A 413 | NP_001276063.1 | |
NM_152332.5 | 1641 | Missense Mutation | CCA,GCA | P,A 477 | NP_689545.1 |