Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGGCCCTTGCCACGACCTACT[C/T]AGGCTACAATTTCCTTTAAGGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616162 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHNYN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KHNYN - KH and NYN domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290256.1 | 873 | UTR 3 | NP_001277185.1 | |||
NM_001290257.1 | 873 | UTR 3 | NP_001277186.1 | |||
NM_015299.2 | 873 | UTR 3 | NP_056114.1 | |||
XM_005267474.4 | 873 | UTR 3 | XP_005267531.1 | |||
XM_011536590.2 | 873 | UTR 3 | XP_011534892.1 | |||
XM_017021114.1 | 873 | Intron | XP_016876603.1 |
LOC101927045 - uncharacterized LOC101927045 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDR39U1 - short chain dehydrogenase/reductase family 39U member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290292.1 | 873 | Silent Mutation | TAA,TGA | *,* 212 | NP_001277221.1 | |
NM_001290293.2 | 873 | Silent Mutation | TAA,TGA | *,* 186 | NP_001277222.1 | |
NM_001308075.1 | 873 | Silent Mutation | TAA,TGA | *,* 212 | NP_001295004.1 | |
NM_020195.2 | 873 | Silent Mutation | TAA,TGA | *,* 294 | NP_064580.2 | |
XM_011536975.1 | 873 | Silent Mutation | TAA,TGA | *,* 317 | XP_011535277.1 | |
XM_011536976.1 | 873 | Silent Mutation | TAA,TGA | *,* 252 | XP_011535278.1 |