Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGCACTCCGTAGCCGCCGCATC[G/T]TTGCCCTGAGCTTGGAGGACCTCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TTC7B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTC7B - tetratricopeptide repeat domain 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010854.1 | 2085 | Silent Mutation | NP_001010854.1 | |||
NM_001320421.1 | 2085 | Silent Mutation | NP_001307350.1 | |||
XM_005267367.1 | 2085 | Silent Mutation | XP_005267424.1 | |||
XM_011536497.1 | 2085 | Silent Mutation | XP_011534799.1 | |||
XM_017021044.1 | 2085 | Silent Mutation | XP_016876533.1 | |||
XM_017021045.1 | 2085 | Silent Mutation | XP_016876534.1 | |||
XM_017021046.1 | 2085 | Silent Mutation | XP_016876535.1 | |||
XM_017021047.1 | 2085 | Silent Mutation | XP_016876536.1 |