Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGCTGCAGTAAGAATGTCTTTT[C/T]CACCTCATTTGAATCGCCCTCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612427 | ||||||||||||||||||||
Literature Links: |
RBM25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM25 - RNA binding motif protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021239.2 | 218 | Missense Mutation | CCA,TCA | P,S 4 | NP_067062.1 | |
XM_011537044.2 | 218 | Missense Mutation | CCA,TCA | P,S 4 | XP_011535346.1 | |
XM_011537045.2 | 218 | UTR 5 | XP_011535347.1 |