Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTGCTCGTCTGTCGTGGAGTCC[C/T]GGCACCAGCCCTCCTGGCGGCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601905 MIM: 190195 | ||||||||||||||||||||
Literature Links: |
RABGGTA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RABGGTA - Rab geranylgeranyltransferase alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004581.5 | 1540 | Missense Mutation | CAG,CGG | Q,R 343 | NP_004572.3 | |
NM_182836.2 | 1540 | Missense Mutation | CAG,CGG | Q,R 343 | NP_878256.1 |
TGM1 - transglutaminase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |