Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATGTCGCTCCCGCGCCAGGAGG[A/G]GAAAAAGGAGCCCCAGCAGCTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608193 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPO4 - importin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024658.3 | 2971 | Missense Mutation | CCC,CTC | P,L 947 | NP_078934.3 |
REC8 - REC8 meiotic recombination protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM9SF1 - transmembrane 9 superfamily member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |