Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCAGTGGCAGAGAATTTCCGAA[C/T]GGAAGAAGTCTATTTACCAGATAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608642 | ||||||||||||||||||||
Literature Links: |
PTGR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGR2 - prostaglandin reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146154.1 | 282 | Missense Mutation | ACG,ATG | T,M 25 | NP_001139626.1 | |
NM_001146155.1 | 282 | Missense Mutation | ACG,ATG | T,M 25 | NP_001139627.1 | |
NM_152444.2 | 282 | Missense Mutation | ACG,ATG | T,M 25 | NP_689657.1 |