Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCAATGGTATATGAGCGGCCTG[C/T]GGGGTTATGAATAGCAGAACAGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604045 | ||||||||||||||||||||
Literature Links: |
LOC101926933 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101926933 - uncharacterized LOC101926933 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRMT5 - protein arginine methyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039619.2 | 1732 | Missense Mutation | ACA,GCA | T,A 612 | NP_001034708.1 | |
NM_001282953.1 | 1732 | Missense Mutation | ACA,GCA | T,A 568 | NP_001269882.1 | |
NM_001282954.1 | 1732 | Missense Mutation | ACA,GCA | T,A 523 | NP_001269883.1 | |
NM_001282955.1 | 1732 | Missense Mutation | ACA,GCA | T,A 585 | NP_001269884.1 | |
NM_001282956.1 | 1732 | Missense Mutation | ACA,GCA | T,A 458 | NP_001269885.1 | |
NM_006109.4 | 1732 | Missense Mutation | ACA,GCA | T,A 629 | NP_006100.2 | |
XM_017020927.1 | 1732 | Missense Mutation | ACA,GCA | T,A 523 | XP_016876416.1 |
PRMT5-AS1 - PRMT5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM23 - RNA binding motif protein 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |