Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGGGGCTTGGCCTCATGGCCCC[C/T]GATGATCTCCTCTGAAAGGAAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116831 | ||||||||||||||||||||
Literature Links: |
GZMH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GZMH - granzyme H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270780.1 | 174 | Missense Mutation | NP_001257709.1 | |||
NM_001270781.1 | 174 | Missense Mutation | NP_001257710.1 | |||
NM_033423.4 | 174 | Missense Mutation | NP_219491.1 | |||
XM_011536683.2 | 174 | UTR 5 | XP_011534985.1 |