Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGGTTGCAGTACCTTGCAGGAA[C/T]GGGAACGTCATTCTGAGGGTAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC254028 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC254028 - uncharacterized LOC254028 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNASE11 - ribonuclease A family member 11 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145250.3 | 221 | Intron | NP_660293.1 |
RNASE12 - ribonuclease A family member 12 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024822.2 | 221 | Missense Mutation | ATT,GTT | I,V 41 | NP_001019993.1 |