Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCCCCCCCAGCCCTGCCTCTCC[A/G]GGTCTGGACAGCCCGGAGCCCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610018 MIM: 605036 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF40 - Rho guanine nucleotide exchange factor 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278529.1 | 2304 | Intron | NP_001265458.1 | |||
NM_001278530.1 | 2304 | Intron | NP_001265459.1 | |||
NM_018071.4 | 2304 | Intron | NP_060541.3 | |||
XM_005267844.2 | 2304 | Intron | XP_005267901.1 | |||
XM_011536937.2 | 2304 | Intron | XP_011535239.1 | |||
XM_011536938.2 | 2304 | Intron | XP_011535240.1 | |||
XM_017021434.1 | 2304 | Intron | XP_016876923.1 | |||
XM_017021435.1 | 2304 | Intron | XP_016876924.1 | |||
XM_017021436.1 | 2304 | Intron | XP_016876925.1 | |||
XM_017021437.1 | 2304 | Intron | XP_016876926.1 |
TMEM253 - transmembrane protein 253 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF219 - zinc finger protein 219 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101672.1 | 2304 | Silent Mutation | NP_001095142.1 | |||
NM_001102454.1 | 2304 | Silent Mutation | NP_001095924.1 | |||
NM_016423.2 | 2304 | Silent Mutation | NP_057507.2 | |||
XM_005267739.1 | 2304 | Silent Mutation | XP_005267796.1 | |||
XM_006720163.2 | 2304 | Silent Mutation | XP_006720226.1 | |||
XM_006720164.3 | 2304 | Silent Mutation | XP_006720227.1 | |||
XM_011536825.1 | 2304 | Silent Mutation | XP_011535127.1 | |||
XM_017021354.1 | 2304 | Silent Mutation | XP_016876843.1 | |||
XM_017021355.1 | 2304 | Silent Mutation | XP_016876844.1 |