Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAAGGCTGAGATGACAGGCCAC[A/G]TGTTCAGCATCAGGCTCACCTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612978 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBLN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBLN3 - cerebellin 3 precursor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039771.2 | 911 | Missense Mutation | ACG,ATG | T,M 147 | NP_001034860.1 |
KHNYN - KH and NYN domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290256.1 | 911 | Intron | NP_001277185.1 | |||
NM_001290257.1 | 911 | Intron | NP_001277186.1 | |||
NM_015299.2 | 911 | Intron | NP_056114.1 | |||
XM_005267474.4 | 911 | Intron | XP_005267531.1 | |||
XM_011536590.2 | 911 | Intron | XP_011534892.1 | |||
XM_017021114.1 | 911 | Intron | XP_016876603.1 |
NYNRIN - NYN domain and retroviral integrase containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |