Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTAAAGGGAAACAGCATGACTG[T/C]GGGGCGCTCCTTATGGCCTCCCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610781 MIM: 603171 MIM: 190195 MIM: 604319 | ||||||||||||||||||||
Literature Links: |
GMPR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMPR2 - guanosine monophosphate reductase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEDD8 - neural precursor cell expressed, developmentally down-regulated 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEDD8-MDP1 - NEDD8-MDP1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGM1 - transglutaminase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TINF2 - TERF1 interacting nuclear factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099274.1 | 724 | Missense Mutation | ACA,GCA | T,A 284 | NP_001092744.1 | |
NM_012461.2 | 724 | Missense Mutation | ACA,GCA | T,A 284 | NP_036593.2 | |
XM_005267529.3 | 724 | Missense Mutation | ACA,GCA | T,A 249 | XP_005267586.1 | |
XM_011536642.2 | 724 | UTR 3 | XP_011534944.1 | |||
XM_017021216.1 | 724 | Missense Mutation | ACA,GCA | T,A 70 | XP_016876705.1 | |
XM_017021217.1 | 724 | Missense Mutation | ACA,GCA | T,A 70 | XP_016876706.1 |