Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGCGCAGTCGCCTGGAGGAGAA[A/G]TTCTTGGAGGAGAACATCTTGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607861 | ||||||||||||||||||||
Literature Links: |
DACT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DACT1 - dishevelled binding antagonist of beta catenin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079520.1 | 470 | Silent Mutation | AAA,AAG | K,K 102 | NP_001072988.1 | |
NM_016651.5 | 470 | Silent Mutation | AAA,AAG | K,K 102 | NP_057735.2 | |
XM_006720167.3 | 470 | Intron | XP_006720230.1 |