Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGTGGCTCAGGAATTAAATTCC[A/C]GAATAGAGACCAGCGATACCTATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605272 MIM: 612484 MIM: 616956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDRG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNASE13 - ribonuclease A family member 13 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012264.3 | 600 | Silent Mutation | NP_001012264.1 |
RNASE7 - ribonuclease A family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPPP2 - tubulin polymerization promoting protein family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173846.4 | 600 | Intron | NP_776245.2 | |||
XM_005267324.4 | 600 | Intron | XP_005267381.1 | |||
XM_011536416.1 | 600 | Intron | XP_011534718.1 | |||
XM_011536417.2 | 600 | Intron | XP_011534719.1 | |||
XM_011536418.1 | 600 | Intron | XP_011534720.1 | |||
XM_011536420.2 | 600 | Intron | XP_011534722.1 | |||
XM_017020966.1 | 600 | Intron | XP_016876455.1 |