Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGGAATCTGAATCTAAGTCAG[C/G]CTTTGGATCCATTTCCATCTCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609798 | ||||||||||||||||||||
Literature Links: |
NEK9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NEK9 - NIMA related kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005268208.1 | 2852 | Missense Mutation | CCT,GCT | P,A 972 | XP_005268265.1 | |
XM_005268209.3 | 2852 | Missense Mutation | CCT,GCT | P,A 854 | XP_005268266.1 | |
XM_017021765.1 | 2852 | Missense Mutation | CCT,GCT | P,A 842 | XP_016877254.1 |
ZC2HC1C - zinc finger C2HC-type containing 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |