Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAATGAACTACGACTCCCAGCAG[C/T]CCCCGTTGCCGCCGCTGCCGTACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616844 MIM: 613256 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC17 - DnaJ heat shock protein family (Hsp40) member C17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018163.2 | 536 | Intron | NP_060633.1 | |||
XM_017022401.1 | 536 | Intron | XP_016877890.1 | |||
XM_017022402.1 | 536 | Intron | XP_016877891.1 |
PPP1R14D - protein phosphatase 1 regulatory inhibitor subunit 14D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFYVE19 - zinc finger FYVE-type containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077268.1 | 536 | Missense Mutation | CCC,TCC | P,S 8 | NP_001070736.1 | |
NM_001258420.1 | 536 | Missense Mutation | CCC,TCC | P,S 8 | NP_001245349.1 | |
NM_001258421.1 | 536 | UTR 5 | NP_001245350.1 | |||
NM_032850.4 | 536 | Intron | NP_116239.3 | |||
XM_017022683.1 | 536 | Intron | XP_016878172.1 | |||
XM_017022684.1 | 536 | Missense Mutation | CCC,TCC | P,S 8 | XP_016878173.1 |