Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACTTCTCTGGACTCAGTTCCTG[C/G]GGCTGTGGGGGTCTTAGGCACTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147521 MIM: 151520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPKA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITPKA - inositol-trisphosphate 3-kinase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002220.2 | 2557 | Intron | NP_002211.1 | |||
XM_011521522.2 | 2557 | Intron | XP_011519824.1 |
LTK - leukocyte receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135685.1 | 2557 | Silent Mutation | CCC,CCG | P,P 690 | NP_001129157.1 | |
NM_002344.5 | 2557 | Silent Mutation | CCC,CCG | P,P 820 | NP_002335.2 | |
NM_206961.3 | 2557 | Silent Mutation | CCC,CCG | P,P 759 | NP_996844.1 | |
XM_011521556.2 | 2557 | Silent Mutation | CCC,CCG | P,P 836 | XP_011519858.1 | |
XM_011521557.2 | 2557 | Silent Mutation | CCC,CCG | P,P 780 | XP_011519859.1 | |
XM_017022181.1 | 2557 | Silent Mutation | CCC,CCG | P,P 841 | XP_016877670.1 | |
XM_017022182.1 | 2557 | Silent Mutation | CCC,CCG | P,P 793 | XP_016877671.1 | |
XM_017022183.1 | 2557 | Silent Mutation | CCC,CCG | P,P 756 | XP_016877672.1 |