Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAGCAAAAATCAGAGGAAGAGCT[C/G]AAGGACAAGAACCAGCTGTTAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614718 | ||||||||||||||||||||
Literature Links: |
KNSTRN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KNSTRN - kinetochore localized astrin/SPAG5 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142761.1 | 640 | Silent Mutation | CTC,CTG | L,L 175 | NP_001136233.1 | |
NM_001142762.1 | 640 | Silent Mutation | CTC,CTG | L,L 175 | NP_001136234.1 | |
NM_033286.3 | 640 | Silent Mutation | CTC,CTG | L,L 175 | NP_150628.3 |