Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACTGGATTCCTGTGGCTGCGTG[C/T]AGGCATTCTCGTTCCACCAATCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610835 | ||||||||||||||||||||
Literature Links: |
ICE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ICE2 - interactor of little elongation complex ELL subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018089.2 | 2954 | Missense Mutation | ACA,GCA | T,A 811 | NP_001018099.1 | |
NM_001276385.1 | 2954 | Intron | NP_001263314.1 | |||
NM_024611.5 | 2954 | Missense Mutation | ACA,GCA | T,A 948 | NP_078887.2 | |
XM_005254661.1 | 2954 | Missense Mutation | ACA,GCA | T,A 948 | XP_005254718.1 | |
XM_005254662.1 | 2954 | Missense Mutation | ACA,GCA | T,A 811 | XP_005254719.1 | |
XM_011522009.1 | 2954 | Missense Mutation | ACA,GCA | T,A 948 | XP_011520311.1 | |
XM_011522010.1 | 2954 | Missense Mutation | ACA,GCA | T,A 811 | XP_011520312.1 | |
XM_011522011.2 | 2954 | Missense Mutation | ACA,GCA | T,A 811 | XP_011520313.1 | |
XM_017022569.1 | 2954 | Missense Mutation | ACA,GCA | T,A 948 | XP_016878058.1 |