Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGTTGAAGGTGAGGGCTACTC[C/T]CACATTGCTCTTCATGATTCTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603489 | ||||||||||||||||||||
Literature Links: |
CILP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CILP - cartilage intermediate layer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003613.3 | 3515 | Missense Mutation | AGA,GGA | R,G 1112 | NP_003604.3 | |
XM_017022678.1 | 3515 | Missense Mutation | AGA,GGA | R,G 1139 | XP_016878167.1 | |
XM_017022679.1 | 3515 | Missense Mutation | AGA,GGA | R,G 1088 | XP_016878168.1 |