Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGTGCCACCATTCATGAGGCGGT[A/C]GTCAGGGCCAGCCCCTGGCCGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C15orf59 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C15orf59 - chromosome 15 open reading frame 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039614.2 | 643 | Missense Mutation | GAC,TAC | D,Y 139 | NP_001034703.1 | |
NM_001303254.1 | 643 | Missense Mutation | GAC,TAC | D,Y 111 | NP_001290183.1 | |
XM_005254369.4 | 643 | Missense Mutation | GAC,TAC | D,Y 139 | XP_005254426.1 |