Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGAGGACAGGCTCAGAAGTTC[C/T]CATAGTGGTGCATGAAGTGGGCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606759 | ||||||||||||||||||||
Literature Links: |
DUOX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUOX2 - dual oxidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014080.4 | 4840 | Missense Mutation | GAG,GGG | E,G 1546 | NP_054799.4 | |
XM_005254421.2 | 4840 | Missense Mutation | GAG,GGG | E,G 1546 | XP_005254478.1 |