Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAGTATCTAAACAGCCAGGCCC[G/T]GTGACAAATGGTCAGCTTCAGCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602534 | ||||||||||||||||||||
Literature Links: |
LRRC57 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC57 - leucine rich repeat containing 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153260.2 | 864 | Intron | NP_694992.2 | |||
XM_011521423.2 | 864 | UTR 3 | XP_011519725.1 | |||
XM_011521424.2 | 864 | Intron | XP_011519726.1 |
SNAP23 - synaptosome associated protein 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003825.3 | 864 | Silent Mutation | CCG,CCT | P,P 121 | NP_003816.2 | |
NM_130798.2 | 864 | Intron | NP_570710.1 | |||
XM_006720725.2 | 864 | Silent Mutation | CCG,CCT | P,P 132 | XP_006720788.1 | |
XM_017022692.1 | 864 | Silent Mutation | CCG,CCT | P,P 132 | XP_016878181.1 | |
XM_017022693.1 | 864 | Intron | XP_016878182.1 | |||
XM_017022694.1 | 864 | Intron | XP_016878183.1 |