Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGACGCTCACGGCTCCGGGGGC[C/T]CATGGCCAGAAGAGACTCAGGAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611246 | ||||||||||||||||||||
Literature Links: |
ADAL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAL - adenosine deaminase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012969.3 | 137 | UTR 5 | NP_001012987.1 | |||
NM_001159280.2 | 137 | UTR 5 | NP_001152752.1 | |||
NM_001324364.1 | 137 | UTR 5 | NP_001311293.1 | |||
NM_001324365.1 | 137 | UTR 5 | NP_001311294.1 | |||
NM_001324366.1 | 137 | UTR 5 | NP_001311295.1 | |||
NM_001324367.1 | 137 | UTR 5 | NP_001311296.1 | |||
NM_001324368.1 | 137 | UTR 5 | NP_001311297.1 | |||
XM_011521291.2 | 137 | Intron | XP_011519593.1 | |||
XM_017021964.1 | 137 | Intron | XP_016877453.1 | |||
XM_017021965.1 | 137 | Intron | XP_016877454.1 | |||
XM_017021966.1 | 137 | Intron | XP_016877455.1 | |||
XM_017021967.1 | 137 | Intron | XP_016877456.1 | |||
XM_017021968.1 | 137 | Intron | XP_016877457.1 | |||
XM_017021969.1 | 137 | Intron | XP_016877458.1 |
LCMT2 - leucine carboxyl methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014793.4 | 137 | Missense Mutation | AGC,GGC | S,G 2 | NP_055608.2 |