Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTTCTTTCTCTCAGAGCTCTGC[A/G]TGCCTCTTGCTGTGCCCTACCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616786 MIM: 606088 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JMJD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JMJD7 - jumonji domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114632.1 | 113 | Missense Mutation | ATG,GTG | M,V 25 | NP_001108104.1 |
JMJD7-PLA2G4B - JMJD7-PLA2G4B readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198588.1 | 113 | Missense Mutation | ATG,GTG | M,V 25 | NP_001185517.1 | |
NM_005090.3 | 113 | Missense Mutation | ATG,GTG | M,V 25 | NP_005081.1 |
MAPKBP1 - mitogen-activated protein kinase binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G4B - phospholipase A2 group IVB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |