Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCAAAGCACCGGCCGAGCGCGA[C/T]CTCCTTGGCCGTGCGAGGGTCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603489 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CILP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CILP - cartilage intermediate layer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003613.3 | 3449 | Missense Mutation | ATC,GTC | I,V 1090 | NP_003604.3 | |
XM_017022678.1 | 3449 | Missense Mutation | ATC,GTC | I,V 1117 | XP_016878167.1 | |
XM_017022679.1 | 3449 | Missense Mutation | ATC,GTC | I,V 1066 | XP_016878168.1 |