Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCTCAGGCAGCCACTCCAGAG[A/G]CGAGAGGGGCATCCAGGTCTCCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608689 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MESP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MESP1 - mesoderm posterior bHLH transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018670.3 | 853 | Missense Mutation | CCT,TCT | P,S 259 | NP_061140.1 |
WDR93 - WD repeat domain 93 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |