Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCATTGTCAAAGTCAAGACAGAA[G/T]GGGGGTCAGCTGAGCCCTCTCAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606471 MIM: 608963 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOP10 - NOP10 ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUTM1 - NUT midline carcinoma family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284292.1 | 752 | Missense Mutation | GGG,TGG | G,W 124 | NP_001271221.1 | |
NM_001284293.1 | 752 | Missense Mutation | GGG,TGG | G,W 114 | NP_001271222.1 | |
NM_175741.2 | 752 | Missense Mutation | GGG,TGG | G,W 96 | NP_786883.1 |