Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGCCCTGGGAGCCTAGGGTAC[A/C]AAGGGTACCGAGGAAGCCATGGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605381 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371031 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371031 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017022800.1 | 1500 | Intron | XP_016878289.1 |
RHCG - Rh family C glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321041.1 | 1500 | Missense Mutation | TTG,TTT | L,F 477 | NP_001307970.1 | |
NM_016321.2 | 1500 | Missense Mutation | TTG,TTT | L,F 477 | NP_057405.1 |