Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCAGTGTCCTGAATCGACTCTG[C/T]CGGCTCGGCACAGACTATATTCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609610 | ||||||||||||||||||||
Literature Links: |
TUBGCP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TUBGCP4 - tubulin gamma complex associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286414.2 | 434 | Silent Mutation | TGC,TGT | C,C 46 | NP_001273343.1 | |
NM_014444.4 | 434 | Silent Mutation | TGC,TGT | C,C 46 | NP_055259.2 | |
XM_011521454.2 | 434 | Silent Mutation | TGC,TGT | C,C 46 | XP_011519756.1 | |
XM_011521455.2 | 434 | UTR 5 | XP_011519757.1 | |||
XM_017022078.1 | 434 | Silent Mutation | TGC,TGT | C,C 46 | XP_016877567.1 |
ZSCAN29 - zinc finger and SCAN domain containing 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |