Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAACAAGCTACAACCCAAGAGAA[C/T]GGCAGATGCGATGAATCTCAGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFAP1 - microfibrillar associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR76 - WD repeat domain 76 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167941.1 | 392 | Missense Mutation | ACG,ATG | T,M 64 | NP_001161413.1 | |
NM_024908.3 | 392 | Missense Mutation | ACG,ATG | T,M 128 | NP_079184.2 | |
XM_005254680.3 | 392 | Missense Mutation | ACG,ATG | T,M 64 | XP_005254737.1 |