Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCACAGCATTTGGAAGTCACTGG[A/G]TCAGAGCCTGTATCACATCCTTTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602166 | ||||||||||||||||||||
Literature Links: |
AP3B2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP3B2 - adaptor related protein complex 3 beta 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278511.1 | 3501 | Missense Mutation | NP_001265440.1 | |||
NM_001278512.1 | 3501 | Missense Mutation | NP_001265441.1 | |||
NM_004644.4 | 3501 | Missense Mutation | NP_004635.2 | |||
XM_017022640.1 | 3501 | Missense Mutation | XP_016878129.1 | |||
XM_017022641.1 | 3501 | Intron | XP_016878130.1 |
CPEB1-AS1 - CPEB1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |