Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCTGGAGGAGGAGCTGCGCGGC[C/T]GGCGCGGGCGAGAGGGCCTGTGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606087 | ||||||||||||||||||||
Literature Links: |
SYNM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYNM - synemin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015286.5 | 256 | Missense Mutation | CGG,TGG | R,W 46 | NP_056101.5 | |
NM_145728.2 | 256 | Missense Mutation | CGG,TGG | R,W 46 | NP_663780.2 | |
XM_017022035.1 | 256 | Missense Mutation | CGG,TGG | R,W 46 | XP_016877524.1 |