Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACTCCGCAGCCTCTCCATCTGC[C/T]GGCACTCCAGCCGCGTGCAGGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609088 MIM: 604728 | ||||||||||||||||||||
Literature Links: |
FBXL22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXL22 - F-box and leucine rich repeat protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203373.2 | 234 | Missense Mutation | CGG,TGG | R,W 78 | NP_976307.2 | |
XM_005254320.3 | 234 | Missense Mutation | CGG,TGG | R,W 78 | XP_005254377.1 | |
XM_006720478.1 | 234 | UTR 5 | XP_006720541.1 | |||
XM_011521468.2 | 234 | UTR 5 | XP_011519770.1 | |||
XM_011521469.1 | 234 | UTR 5 | XP_011519771.1 | |||
XM_011521472.2 | 234 | UTR 5 | XP_011519774.1 | |||
XM_017022086.1 | 234 | Missense Mutation | CGG,TGG | R,W 78 | XP_016877575.1 |
USP3 - ubiquitin specific peptidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP3-AS1 - USP3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |