Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGGAGACGGCGACTTCTGACAA[A/C]GAGTAAGCGCCTCATTGATCTCGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609291 | ||||||||||||||||||||
Literature Links: |
SPRED1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPRED1 - sprouty related EVH1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152594.2 | 1129 | Missense Mutation | AAA,AAC | K,N 10 | NP_689807.1 | |
XM_005254202.3 | 1129 | Missense Mutation | AAA,AAC | K,N 10 | XP_005254259.1 | |
XM_011521289.2 | 1129 | Intron | XP_011519591.1 |