Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCTCTTCCCAGCAGGCTGGCC[A/G]GGAGCCCGCGCGATAGTGGAGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147370 | ||||||||||||||||||||
Literature Links: |
IGF1R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGF1R - insulin like growth factor 1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGPEP1L - pyroglutamyl-peptidase I-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102612.2 | 765 | Missense Mutation | CCG,CTG | P,L 155 | NP_001096082.2 | |
NM_001167902.1 | 765 | Intron | NP_001161374.1 | |||
XM_011521254.1 | 765 | Missense Mutation | CCG,CTG | P,L 246 | XP_011519556.1 | |
XM_017021933.1 | 765 | Missense Mutation | CCG,CTG | P,L 101 | XP_016877422.1 |